Anti-IL6R Rabbit Polyclonal Antibody
Lieferant: US Biological
I8428-39M3
USBII8428-39M3EA 633 EUR
USBII8428-39M3
Anti-IL6R Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:40000; Western Blot: 1:200-1:500; Immunochemistry: 2.5 ?g/ml
Type: Primary
Antigen: IL6R (interleukin 6 receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human