Anti-Iron Responsive Element Binding Protein 2 Mouse Monoclonal Antibody [clone: 10E57]

Lieferant: US Biological
I8615-75D
USBII8615-75DEA 553 EUR
USBII8615-75D
Anti-Iron Responsive Element Binding Protein 2 Mouse Monoclonal Antibody [clone: 10E57]
Antikörper
Type: Primary
Antigen: Iron Responsive Element Binding Protein 2
Clonality: monoclonal
Clone: 1E+58
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR