Anti-IDH1 Goat Polyclonal Antibody

Lieferant: US Biological
I8850-40C
USBII8850-40CEA 640 EUR
USBII8850-40C
Anti-IDH1 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 0.1-0. 3?g/ml

Type: Primary
Antigen: IDH1 (isocitrate dehydrogenase 1 (NADP+), soluble)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Rat, Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR