Anti-J Chain Goat Polyclonal Antibody
Lieferant: US Biological
J1255-01
USBIJ1255-01EA 608 EUR
USBIJ1255-01
Anti-J Chain Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: J Chain
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human