Anti-MAPK8 Rabbit Polyclonal Antibody

Lieferant: US Biological
J2475-03C
USBIJ2475-03CEA 489 EUR
USBIJ2475-03C
Anti-MAPK8 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 1:50-1:100

Type: Primary
Antigen: MAPK8 (mitogen-activated protein kinase 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR