Anti-PSA Rabbit Polyclonal Antibody

Lieferant: US Biological
K0005-06A
USBIK0005-06AEA 505 EUR
USBIK0005-06A
Anti-PSA Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:500-1:1000

Type: Primary
Antigen: PSA
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR