Anti-KLH Mouse Monoclonal Antibody

Lieferant: US Biological
K0300-03D
USBIK0300-03DEA 601 EUR
USBIK0300-03D
Anti-KLH Mouse Monoclonal Antibody
Antikörper
Recommended dilution: ELISA: 0.5-2?g/ml

Type: Tag
Antigen: KLH (Keyhole Limpet Hemocyanin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR