Anti-KIF5B Mouse Monoclonal Antibody [clone: KN-03]

Lieferant: US Biological
K1759-24H
USBIK1759-24HEA 647 EUR
USBIK1759-24H
Anti-KIF5B Mouse Monoclonal Antibody [clone: KN-03]
Antikörper
Recommended dilution: Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: KIF5B (kinesin family member 5B)
Clonality: monoclonal
Clone: KN-03
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR