Anti-Kir, Low Endotoxin Mouse Monoclonal Antibody [clone: 4j52]

Lieferant: US Biological
K1853-04
USBIK1853-04EA 903 EUR
USBIK1853-04
Anti-Kir, Low Endotoxin Mouse Monoclonal Antibody [clone: 4j52]
Antikörper
Type: Primary
Antigen: Kir, Low Endotoxin
Clonality: monoclonal
Clone: 4j52
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR