Anti-KT3 Rabbit Polyclonal Antibody

Lieferant: US Biological
K6980-05
USBIK6980-05EA 337 EUR
USBIK6980-05
Anti-KT3 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 0. 05-0.2?g/ml; Western Blot: 0.1-1.0 ?g/ml

Type: Tag
Antigen: KT3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR