Anti-L-Kynurenine Rabbit Polyclonal Antibody (Unconjugated)
Lieferant: US Biological
K9799-90E
USBIK9799-90EEA 890 EUR
USBIK9799-90E
Anti-L-Kynurenine Rabbit Polyclonal Antibody (Unconjugated)
Antikörper
Type: Primary
Antigen: L-Kynurenine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen