Anti-Kynurenine 3-monooxygenase Rabbit Polyclonal Antibody

Lieferant: US Biological
K9800-40
USBIK9800-40EA 649 EUR
USBIK9800-40
Anti-Kynurenine 3-monooxygenase Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: Kynurenine 3-monooxygenase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR