Anti-Kynurenine 3-monooxygenase Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
K9800-40-BIOT
USBIK9800-40-BIOTEA 884 EUR
USBIK9800-40-BIOT
Anti-Kynurenine 3-monooxygenase Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Kynurenine 3-monooxygenase
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR