Anti-Kynurenine 3-Monooxygenase Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: US Biological
K9800-40-HRP
USBIK9800-40-HRPEA 884 EUR
USBIK9800-40-HRP
Anti-Kynurenine 3-Monooxygenase Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Type: Primary
Antigen: Kynurenine 3-monooxygenase
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human