Anti-LAG3 Mouse Monoclonal Antibody ATTO 647 [clone: 6D65]

Lieferant: US Biological
L1050-61C
USBIL1050-61CEA 736 EUR
USBIL1050-61C
Anti-LAG3 Mouse Monoclonal Antibody ATTO 647 [clone: 6D65]
Antikörper
Type: Primary
Antigen: LAG3 (lymphocyte-activation gene 3)
Clonality: monoclonal
Clone: 6D65
Conjugation: ATTO 647
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR