Anti-lambda light chain Goat Polyclonal Antibody

Lieferant: US Biological
L1185-11L
USBIL1185-11LEA 298 EUR
USBIL1185-11L
Anti-lambda light chain Goat Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: LAMBDA LIGHT CHAIN
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR