Anti-LAT Mouse Monoclonal Antibody [clone: LAT-01]

Lieferant: US Biological
L1390-01J
USBIL1390-01JEA 742 EUR
USBIL1390-01J
Anti-LAT Mouse Monoclonal Antibody [clone: LAT-01]
Antikörper
Type: Primary
Antigen: LAT (linker for activation of T cells)
Clonality: monoclonal
Clone: LAT-01
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR