Anti-LAT Goat Polyclonal Antibody

Lieferant: US Biological
L1390-05A
USBIL1390-05AEA 601 EUR
USBIL1390-05A
Anti-LAT Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LAT (linker for activation of T cells)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR