Anti-LEP Mouse Monoclonal Antibody [clone: 7C216]

Lieferant: US Biological
L1670-08P
USBIL1670-08PEA 532 EUR
USBIL1670-08P
Anti-LEP Mouse Monoclonal Antibody [clone: 7C216]
Antikörper
Recommended dilution: Immunofluorescence: 1:1000; Western Blot: 1:4000

Type: Primary
Antigen: LEP (leptin)
Clonality: monoclonal
Clone: 7C216
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR