Anti-Leukocyte Immunoglobulin-Like Receptor C2 Sheep Polyclonal Antibody (Biotin)

Lieferant: US Biological
L2028-16
USBIL2028-16EA 910 EUR
USBIL2028-16
Anti-Leukocyte Immunoglobulin-Like Receptor C2 Sheep Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: Western Blot: 0.1-0.2?g/ml

Type: Primary
Antigen: Leukocyte Immunoglobulin-Like Receptor C2
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR