Anti-LCN2 Mouse Monoclonal Antibody [clone: 13A129]
Lieferant: US Biological
L2499-14C
USBIL2499-14CEA 1170 EUR
USBIL2499-14C
Anti-LCN2 Mouse Monoclonal Antibody [clone: 13A129]
Antikörper
Type: Primary
Antigen: LCN2 (lipocalin 2)
Clonality: monoclonal
Clone: 13A129
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human