Anti-LCN2 Mouse Monoclonal Antibody [clone: 13A130]

Lieferant: US Biological
L2499-14D
USBIL2499-14DEA 1140 EUR
USBIL2499-14D
Anti-LCN2 Mouse Monoclonal Antibody [clone: 13A130]
Antikörper
Recommended dilution: C?lture Medium:RPMI 1640 with 2-10% fetal calf serum

Type: Primary
Antigen: LCN2 (lipocalin 2)
Clonality: monoclonal
Clone: 13A130
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR