Anti-LCN2 Rat Monoclonal Antibody [clone: 13A131]

Lieferant: US Biological
L2499-14E
USBIL2499-14EEA 1140 EUR
USBIL2499-14E
Anti-LCN2 Rat Monoclonal Antibody [clone: 13A131]
Antikörper
Type: Primary
Antigen: LCN2 (lipocalin 2)
Clonality: monoclonal
Clone: 13A131
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1 Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR