Anti-LCN2 Mouse Monoclonal Antibody [clone: 13A133]
Lieferant: US Biological
L2499-14G
USBIL2499-14GEA 1140 EUR
USBIL2499-14G
Anti-LCN2 Mouse Monoclonal Antibody [clone: 13A133]
Antikörper
Type: Primary
Antigen: LCN2 (lipocalin 2)
Clonality: monoclonal
Clone: 13A133
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Dog