Anti-LCN2 Mouse Monoclonal Antibody [clone: 6F155]
Lieferant: US Biological
L2499-16
USBIL2499-16EA 898 EUR
USBIL2499-16
Anti-LCN2 Mouse Monoclonal Antibody [clone: 6F155]
Antikörper
Type: Primary
Antigen: LCN2 (lipocalin 2)
Clonality: monoclonal
Clone: 6F155
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human