Anti-LCN2 Mouse Monoclonal Antibody [clone: 9G211]

Lieferant: US Biological
L2499-16A
USBIL2499-16AEA 910 EUR
USBIL2499-16A
Anti-LCN2 Mouse Monoclonal Antibody [clone: 9G211]
Antikörper
Recommended dilution: Immunofluorescence: 1:50

Type: Primary
Antigen: LCN2 (lipocalin 2)
Clonality: monoclonal
Clone: 9G211
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR