Anti-LCN2 Goat Polyclonal Antibody
Lieferant: US Biological
L2499-17K
USBIL2499-17KEA 601 EUR
USBIL2499-17K
Anti-LCN2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:8000
Type: Primary
Antigen: LCN2 (lipocalin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse