Anti-LCN2 Goat Polyclonal Antibody

Lieferant: US Biological
L2499-17Q
USBIL2499-17QEA 633 EUR
USBIL2499-17Q
Anti-LCN2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:16000; Western Blot: 1:16000

Type: Primary
Antigen: LCN2 (lipocalin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR