Anti-LCN2 Rabbit Polyclonal Antibody

Lieferant: US Biological
L2499-17U
USBIL2499-17UEA 450 EUR
USBIL2499-17U
Anti-LCN2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: LCN2 (lipocalin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR