Anti-LCN2 Goat Polyclonal Antibody

Lieferant: US Biological
L2499-18
USBIL2499-18EA 897 EUR
USBIL2499-18
Anti-LCN2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 0.2-0. 8?g/ml; Western Blot: 1 ?g/ml

Type: Primary
Antigen: LCN2 (lipocalin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR