Anti-Listeria sp. Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
L2650-09B
USBIL2650-09BEA 459 EUR
USBIL2650-09B
Anti-Listeria sp. Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 1:1000

Type: Primary
Antigen: Listeria sp.
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR