Anti-CAMP Rabbit Polyclonal Antibody

Lieferant: US Biological
L2650-95A
USBIL2650-95AEA 1080 EUR
USBIL2650-95A
Anti-CAMP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CAMP (cathelicidin antimicrobial peptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR