Anti-LLGL2 Rabbit Polyclonal Antibody

Lieferant: US Biological
L2651-75C
USBIL2651-75CEA 649 EUR
USBIL2651-75C
Anti-LLGL2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: LLGL2 (lethal giant larvae homolog 2 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR