Anti-LRRK2 Sheep Polyclonal Antibody
Lieferant: US Biological
L4502-21K
USBIL4502-21KEA 640 EUR
USBIL4502-21K
Anti-LRRK2 Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LRRK2 (leucine-rich repeat kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human