Anti-LUC7-like Rabbit Polyclonal Antibody
Lieferant: US Biological
L5970-01C
USBIL5970-01CEA 649 EUR
USBIL5970-01C
Anti-LUC7-like Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Type: Primary
Antigen: LUC7-like
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human