Anti-Lung Cancer-Associated Antigen Mouse Monoclonal Antibody [clone: 5F266]
Lieferant: US Biological
L6049-73
USBIL6049-73EA 514 EUR
USBIL6049-73
Anti-Lung Cancer-Associated Antigen Mouse Monoclonal Antibody [clone: 5F266]
Antikörper
Recommended dilution: Immunochemistry: 1:5-1:10
Type: Primary
Antigen: Lung Cancer-Associated Antigen
Clonality: monoclonal
Clone: 5F266
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human