Anti-Luteinizing Hormone Mouse Monoclonal Antibody [clone: 9L457]
Lieferant: US Biological
L7500-02J
USBIL7500-02JEA 641 EUR
USBIL7500-02J
Anti-Luteinizing Hormone Mouse Monoclonal Antibody [clone: 9L457]
Antikörper
Type: Primary
Antigen: Luteinizing Hormone
Clonality: monoclonal
Clone: 9L457
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human