Anti-Luteinizing Hormone Mouse Monoclonal Antibody [clone: 9L459]

Lieferant: US Biological
L7500-02L
USBIL7500-02LEA 627 EUR
USBIL7500-02L
Anti-Luteinizing Hormone Mouse Monoclonal Antibody [clone: 9L459]
Antikörper
Type: Primary
Antigen: Luteinizing Hormone
Clonality: monoclonal
Clone: 9L459
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR