Anti-Luteinizing Hormone Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 10g201]

Lieferant: US Biological
L7500-03R
USBIL7500-03REA 627 EUR
USBIL7500-03R
Anti-Luteinizing Hormone Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 10g201]
Antikörper
Recommended dilution: ELISA: 1:1000

Type: Primary
Antigen: Luteinizing Hormone
Clonality: monoclonal
Clone: 10g201
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR