Anti-Luteinizing Hormone Sheep Polyclonal Antibody

Lieferant: US Biological
L7500-04A
USBIL7500-04AEA 479 EUR
USBIL7500-04A
Anti-Luteinizing Hormone Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Luteinizing Hormone
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR