Anti-Luteinizing Hormone Mouse Monoclonal Antibody [clone: 3F255]

Lieferant: US Biological
L7500-05A-250
USBIL7500-05A-250EA 371 EUR
USBIL7500-05A-250
Anti-Luteinizing Hormone Mouse Monoclonal Antibody [clone: 3F255]
Antikörper
Type: Primary
Antigen: Luteinizing Hormone
Clonality: monoclonal
Clone: 3F255
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR