Anti-Luteinizing Hormone, alpha Mouse Monoclonal Antibody [clone: 10g105]

Lieferant: US Biological
L7500-10B
USBIL7500-10BEA 389 EUR
USBIL7500-10B
Anti-Luteinizing Hormone, alpha Mouse Monoclonal Antibody [clone: 10g105]
Antikörper
Type: Primary
Antigen: Luteinizing Hormone, alpha
Clonality: monoclonal
Clone: 10g105
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR