Anti-Luteinizing Hormone Receptor Rabbit Polyclonal Antibody
Lieferant: US Biological
L7509-25C
USBIL7509-25CEA 687 EUR
USBIL7509-25C
Anti-Luteinizing Hormone Receptor Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:100-1:400
Type: Primary
Antigen: Luteinizing Hormone Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat