Anti-Lysophosphatidic Acid Receptor Rabbit Polyclonal Antibody

Lieferant: US Biological
L9189-04C
USBIL9189-04CEA 633 EUR
USBIL9189-04C
Anti-Lysophosphatidic Acid Receptor Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Immunochemistry: 5-10 ?g/ml

Type: Primary
Antigen: Lysophosphatidic Acid Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR