Anti-Lysophosphatidic Acid Receptor Sheep Polyclonal Antibody
Lieferant: US Biological
L9189-04D
USBIL9189-04DEA 817 EUR
USBIL9189-04D
Anti-Lysophosphatidic Acid Receptor Sheep Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml
Type: Primary
Antigen: Lysophosphatidic Acid Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human