Anti-Lysophosphatidic Acid Receptor Rabbit Polyclonal Antibody

Lieferant: US Biological
L9189-05M
USBIL9189-05MEA 444 EUR
USBIL9189-05M
Anti-Lysophosphatidic Acid Receptor Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry; Western Blot: 2.5 ?g/ml

Type: Primary
Antigen: Lysophosphatidic Acid Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR