Anti-Lysophosphatidic Acid Receptor Rabbit Polyclonal Antibody
Lieferant: US Biological
L9189-07A
USBIL9189-07AEA 681 EUR
USBIL9189-07A
Anti-Lysophosphatidic Acid Receptor Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 20 ?g/ml
Type: Primary
Antigen: Lysophosphatidic Acid Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human