Anti-Sphingomyelin phosphodiesterase 1 Goat Polyclonal Antibody

Lieferant: US Biological
L9189-95B
USBIL9189-95BEA 857 EUR
USBIL9189-95B
Anti-Sphingomyelin phosphodiesterase 1 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 0.5-1.0 ?g/ml Western Blot: 1.0 ?g/ml

Type: Primary
Antigen: Sphingomyelin phosphodiesterase 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR