Anti-Monocyte Chemotactic Protein 1 Sheep Polyclonal Antibody

Lieferant: US Biological
M1203-03L
USBIM1203-03LEA 704 EUR
USBIM1203-03L
Anti-Monocyte Chemotactic Protein 1 Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Monocyte Chemotactic Protein 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR