Anti-MAGEL2 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
M1750-15-BIOT
USBIM1750-15-BIOTEA 884 EUR
USBIM1750-15-BIOT
Anti-MAGEL2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: MAGEL2 (MAGE-like 2)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR